Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Water, DEPC Treated, Sterile, Nuclease-Free, OmniPur™, Calbiochem™,

Catalog No. 9602500ML Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
5 L
10 L
Packaging:
Plastic Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
96-025-00ML 500 mL Plastic Bottle
96-011-00ML 100 mL Plastic Bottle
96-101-L 1 L Plastic Bottle
96-125-L 5 L Plastic Bottle
96-131-0L 10 L Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 96-025-00ML Supplier MilliporeSigma™ Supplier No. 9602500ML
Add to Cart
Add to Cart

Used for RNA applications (Prepared with DEPC)

Recommended Storage

+15°C to +30°C.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.015
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym dihydrogen oxide, dihydrogen monoxide
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name oxidane
SMILES O

Specifications

Color Colorless
Grade Molecular Biology
Packaging Plastic Bottle
Quantity 500 mL
Physical Form Liquid
Recommended Storage +15°C to +30°C.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.