Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Water, DEPC-Treated

Catalog No. AAJ62087K2
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ62087K2 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ62087K2 Supplier Thermo Scientific Chemicals Supplier No. J62087K2
  • $150.80
    $128.65 / Each of 1

    Save $22.15

    15% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 7732-18-5 | H2O | 18.015 g/mol

It is used as mobile phase solvent in LC-MS analysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as mobile phase solvent in LC-MS analysis.

Solubility
Soluble in water. Insoluble in polar liquids.

Notes
Suitable for any general chemical storage area. Protect from freezing. Incompatib
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.015
MDL Number MFCD00011332
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name oxidane
SMILES O

Specifications

Color Colorless
Boiling Point 100°C
Beilstein 2050024
Merck Index 14,10039
Solubility Information Soluble in water. Insoluble in polar liquids.
Formula Weight 18.02
Odor Odorless
Density 1.000
Quantity 1 L
Physical Form Liquid
Chemical Name or Material Water, DEPC-Treated
Show More Show Less
EINECSNumber 231-791-2
RTECSNumber ZC0110000
TSCA Yes

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.