Learn More
Growcells SDW-2000-6 Water, Sterile Distilled (Pack of 6) 2L
Catalog No. 501034782
Encompass_Preferred
Sterile Distilled water. Used the preparation of cell culture media and laboratory reagents. Autoclaved in final packaging.
Catalog No. 50-103-4782 Supplier Growcells Supplier No. SDW20006
Sterile Distilled water. Used the preparation of cell culture media and laboratory reagents. Autoclaved in final packaging.