Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Water, DNA Grade, DNASE, Protease free, Fisher BioReagents

Catalog No. BP24701 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
1 L
Packaging:
Nalgene Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BP24701 1 L Nalgene Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BP24701
Add to Cart

CAS: 7732-18-5 | H2O | 18.015 g/mol

Nalgene Poly Bottle; 1L

DNA-grade Water is DNase- and Protease-free and is suitable for all DNA work.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Water
pH 7
Name Note For DNA Work, DNASE, Protease free
Color Colorless
CAS 7732-18-5
Melting Point 0°C
Boiling Point 100°C
Molecular Formula H2O
DNase DNase free
Merck Index 15, 10237
Synonym dihydrogen oxide, dihydrogen monoxide
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
SMILES O
IUPAC Name oxidane
Molecular Weight (g/mol) 18.015
PubChem CID 962
ChEBI CHEBI:15377
Formula Weight 18.02
Grade DNA
Packaging Nalgene Poly Bottle
Vapor Pressure 17.5mmHg at 20°C
Filtered Through Filtered through a 0.2-micron filter and autoclaved.
Density 1.000g/cm³
Quantity 1 L
Physical Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.