Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quality Biological Inc Water, Endotoxin-Free, Case 10 X 500 mL
SDP

Catalog No. NC3662032
Encompass_Preferred

Water, Endotoxin-Free is ideal for preparing solutions and reagents for transfection, transformation, cell and tissue cultures. Use when producing ultra-low, endotoxin serum-free media and buffers. Suitable when testing under pyrogen-free conditions (LAL testing) is required. Reliable for rinsing and washing equipment, glassware, and plasticware in endotoxin-free conditions.

Mycoplasma tested
DNase, RNase and protease tested
Endotoxin < 0.005 EU/mL

Storage Condition: 15 °C to 30 °C
Shipping Condition: ambient temperature
Sterile Filtered 0.1 µm
Endotoxin (<0.005 EU/mL) and Mycoplasma Tested
DNase, RNase, and Protease Tested
Max Shelf Life: 24 months from date of manufacture

Manufactured under an ISO 9001:2015 certified quality management system.

Product and/or packaging contains recycled/renewable content. Product and/or packaging is recyclable or reduces waste. Product and/or packaging is safer to human and environmental health.

Catalog No. NC3662032 Supplier Quality Biological Inc Supplier No. 118325101CS
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Water, Endotoxin-Free is ideal for preparing solutions and reagents for transfection, transformation, cell and tissue cultures. Use when producing ultra-low, endotoxin serum-free media and buffers. Suitable when testing under pyrogen-free conditions (LAL testing) is required. Reliable for rinsing and washing equipment, glassware, and plasticware in endotoxin-free conditions.

Mycoplasma tested
DNase, RNase and protease tested
Endotoxin < 0.005 EU/mL

Storage Condition: 15 °C to 30 °C
Shipping Condition: ambient temperature
Sterile Filtered 0.1 µm
Endotoxin (<0.005 EU/mL) and Mycoplasma Tested
DNase, RNase, and Protease Tested
Max Shelf Life: 24 months from date of manufacture

Manufactured under an ISO 9001:2015 certified quality management system.

Product and/or packaging contains recycled/renewable content. Product and/or packaging is recyclable or reduces waste. Product and/or packaging is safer to human and environmental health.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.