Learn More
Waters Corp Waters Corporation Extraction Manifold, 20-position, no rack
Catalog No. NC2306903 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Waters Extraction Manifold is a 20-position device designed for sample preparation, facilitating efficient extraction processes. This manifold is supplied with essential accessories to support immediate laboratory use.
  • Designed for 20 positions
  • Includes 20 needle tips
  • Includes 25 plugs
  • Includes an ejector tool
  • Operates with a vacuum pump
  • Certificate of analysis available for download
  • Safety data sheet available for download
Catalog No. NC2306903 Supplier Waters Corp Supplier No. 186008998
The Waters Extraction Manifold is a 20-position device designed for sample preparation, facilitating efficient extraction processes. This manifold is supplied with essential accessories to support immediate laboratory use.
  • Designed for 20 positions
  • Includes 20 needle tips
  • Includes 25 plugs
  • Includes an ejector tool
  • Operates with a vacuum pump
  • Certificate of analysis available for download
  • Safety data sheet available for download