Learn More
Lonza Walkersville Water f Cell Culture 200L GMP
Catalog No. 5000496269
Encompass
Water for Cell Culture Water For Injection (WFI) quality water is prepared by ultrafiltration reveresed osmosis deionisaton and sterile filtration Lonza s Water for Cell Culture is aseptically processed from a validated Water for Injection (WFI) system that meets current USP specifications This BioWhittaker Cell Culture Media is produced according to cGMP and labelled For Further Manufacturing (FFM) GMP Products are intended for research or further manufacturing use only THIS PRODUCT IS NOT INTENDED FOR DIRECT THERAPEUTIC USE IN HUMANS
Catalog No. 50-004-96269 Supplier Lonza Walkersville Supplier No. 08199E
Water for Cell Culture Water For Injection (WFI) quality water is prepared by ultrafiltration reveresed osmosis deionisaton and sterile filtration Lonza s Water for Cell Culture is aseptically processed from a validated Water for Injection (WFI) system that meets current USP specifications This BioWhittaker Cell Culture Media is produced according to cGMP and labelled For Further Manufacturing (FFM) GMP Products are intended for research or further manufacturing use only THIS PRODUCT IS NOT INTENDED FOR DIRECT THERAPEUTIC USE IN HUMANS