Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Water, Filtered through a 0.2 micron filter, BAKER ANALYZED™ ULTRA LC/MS, J.T. Baker™

Catalog No. 02004557 Shop All Avantor Products
Change view
Click to view available options
Quantity:
1 L
Packaging:
Narrow Mouth Flint
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02-004-557 1 L Narrow Mouth Flint
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02-004-557 Supplier Avantor Supplier No. 982302
Add to Cart
Add to Cart

For cutting-edge LC/MS applications

    For Use in LC/MS and UHPLC; For Laboratory, Research, or Manufacturing Use

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.015
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name oxidane
SMILES O

Specifications

Formula Weight 18
Grade BAKER ANALYZED™ ULTRA LC/MS
Packaging Narrow Mouth Flint
Quantity 1 L
Specific Gravity 1L = 1.00kg
Chemical Name or Material Water, Filtered through a 0.2 micron filter
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.