Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Fine Chemicals Biosciences Water | 7732-18-5 | MFCD00011332 | 1 L

Catalog No. 501809593
Encompass

Water | Mol Wt: 18.02 | 7732-18-5 | MFCD00011332 | 1 L

Catalog No. 50-180-9593 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 952891L
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Water | Mol Wt: 18.02 | 7732-18-5 | MFCD00011332 | 1 L

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.