Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Water Hardness Buffer, LabChem™

Catalog No. LC268402 Shop All LabChem Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC268402 1 L
LC268401 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LC268402 Supplier LabChem, Inc. Supplier No. LC268402
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Weight: 381.37 g/mol

Specifications

Chemical Name or Material Water Hardness Buffer
Color Colorless
CAS Max % 93.25, 4, 1, 1, 0.65
CAS 1303-96-4, 6381-59-5, 1313-84-4, 1310-73-2
Decomposition Information Carbon monoxide; Carbon dioxide; Sulfur compounds
Physical Form Liquid
Quantity 1 L
Identification Passes Test
Linear Formula Na2B4O7·10H2O
Packaging Poly Bottle
Solubility Information Soluble in water
Formula Weight 381.37
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.