Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Water Hardness Buffer, with Magnesium, Certified, LabChem™

Catalog No. LC268104
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC268104 4 L
LC268101 500 mL
LC268102 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. LC268104 Supplier LabChem, Inc. Supplier No. LC268104
Add to Cart
Add to Cart

Specifications

Chemical Name or Material Water Hardness Buffer
Name Note with Magnesium
Color Colorless
CAS Max % 88.1, 6.8, 4.6, 0.5
CAS 12125-02-9, 1336-21-6, 14402-88-1
Decomposition Information Fume; Carbon monoxide; Carbon dioxide; Thermal decomposition generates: Corrosive vapors
Density 0.96g/mL
Physical Form Liquid
Quantity 4 L
Identification Passes Test
Linear Formula NH4OH
Packaging Poly Bottle
UN Number UN2672
Specific Gravity 0.96g/mL
Solubility Information Soluble in water
Formula Weight 35.05
Grade Certified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.