Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Water Hardness Buffer with Magnesium EDTA, Reagents

Catalog No. 01333392
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-333-392 500 mL
01-333-395 1 L
01-333-394 20 L
01-333-393 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 01-333-392 Supplier Reagents Holdings Llc Supplier No. CW000600500A
Only null left
Add to Cart
Add to Cart

Water Hardness Buffer with Magnesium EDTA, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Color Colorless
Volume (Metric) 500 mL
For Use With (Application) Water Analysis
Packaging Poly Bottle
Quantity 500 mL
Sterility Non-sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.