Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Water Hardness Indicator, 0.5% (w/v) Eriochrome Black T in 2-Methoxyethanol, Ricca Chemical
SDP

Catalog No. 93004 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
120 mL
500 mL
1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
9300-4 120 mL
9300-16 500 mL
930032 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 9300-4 Supplier Ricca Chemical Company Supplier No. 93004
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

APHA/EPA for Water Hardness Testing

Specifications

pH Range Alkaline
Volume (Metric) 120 mL
Packaging Amber Glass Bottle
Quantity 120 mL
Test Method APHA/EPA for Water Hardness
Type Water Hardness Indicator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.