Learn More
Hach Company Water Hardness TNTplus Vial Test (20 - 350 mg/L as CaCO₃) | 25 tests
Catalog No. 501998154 Shop All Hach Company Products
Encompass
Water Hardness TNTplus Vial Test (20 - 350 mg/L as CaCO₃) | 25 tests
Catalog No. 50-199-8154 Supplier Hach Company Supplier No. TNT869
Water Hardness TNTplus Vial Test (20 - 350 mg/L as CaCO₃) | 25 tests