Learn More
Research Products International Corp Water, HPLC Grade, 4 Liter Bottle
SDP
Catalog No. 501367979
Encompass_Preferred
Certified RNase and DNase free for use in molecular biology applications. Ideal for making reagents, rinsing glass and plastic ware, enzymatic reactions and more. Toxic agents such as DEPC are not used in purification procedures.
  • Endotoxin Free
  • RNase Free
  • DNase Free
*Tested to < 0.001 EU/ML
Catalog No. 50-136-7979 Supplier Research Products International Corp Supplier No. 248797
Certified RNase and DNase free for use in molecular biology applications. Ideal for making reagents, rinsing glass and plastic ware, enzymatic reactions and more. Toxic agents such as DEPC are not used in purification procedures.
  • Endotoxin Free
  • RNase Free
  • DNase Free
*Tested to < 0.001 EU/ML