Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Fine Chemicals Biosciences Water HPLC Plus | 7732-18-5 | MFCD00011332 | 4L

Catalog No. 501809579
Encompass

Water HPLC Plus | Mol Wt: 18.02 | 7732-18-5 | MFCD00011332 | 4L

Catalog No. 50-180-9579 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 348774L
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Water HPLC Plus | Mol Wt: 18.02 | 7732-18-5 | MFCD00011332 | 4L

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.