Learn More
Sigma Aldrich Fine Chemicals Biosciences Water HPLC Plus1L
Catalog No. 502898653
Encompass
HPLC Plus grade water is useful for the UHPLC (Ultra-High Pressure Liquid Chromatography) gradient elution and HPLC studies.
Catalog No. 50-289-8653 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 348771L
HPLC Plus grade water is useful for the UHPLC (Ultra-High Pressure Liquid Chromatography) gradient elution and HPLC studies.