Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Qorpak™ Water for Injection

Catalog No. 01000938
Change view
Click to view available options
Density:
20 L
200 L
Packaging:
Bioprocess Bag
Plastic Drum
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Density Packaging
01-000-938 20 L Bioprocess Bag
01-001-033 200 L Plastic Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-000-938 Supplier Qorpak™ Supplier No. 336645
Add to Cart
Add to Cart
Request a Quote

WFI Quality Water is made in a USA state-of-the-art production facility, including ISO 7 Cleanrooms and ISO 5 automated filling stations following cGMP guidelines. The WFI water is stored and shipped in BioProcess Containers (BPCs).

  • Meets or exceeds USP standards and monograph requirements
  • FDA-registered cGMP manufacturing facility
  • U.S. Pharmacopeia (USP<1231>) and ISO 13485 compliant
  • The only water treatment system that has been tested and certified to the new IGC376 standard

Specifications

Chemical Name or Material H2O
Molecular Weight (g/mol) 18.015
Grade USP
Packaging Bioprocess Bag
Sterility Non-sterile
Density 20 L
Quantity 30/Pallet
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.