Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Hydroxide Solution 40% (w/w), Biotech Reagent, J.T. Baker™

Catalog No. 02004099 Shop All Avantor Products
Change view
Click to view available options
Quantity:
19 L
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02-004-099 19 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 02-004-099 Supplier Avantor Supplier No. 089607
Add to Cart
Add to Cart
This item is not returnable. View return policy

Produced under GMP guidelines, but for which there is no USP/NF compendia for the product

Chemical Identifiers

CAS 1310-73-2
Molecular Formula HNaO
Molecular Weight (g/mol) 39.997
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]

Specifications

Quantity 19 L
Formula Weight 40
Percent Purity 40% (w/w)
Grade Biotech Reagent
Packaging Poly Bottle
Specific Gravity 1L = 1.43kg
Chemical Name or Material Water for Injection Quality Water
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.