Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Moore Medical Water-Jel™ Cool Jel

Catalog No. 19898204
Change view
Click to view available options
Volume (Metric):
3.5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
19-898-204 3.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-898-204 Supplier Moore Medical Supplier No. 52107

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Product Type Cool Jel
Color White
For Use With (Application) Comforts and moisturizes the injured area while forming a protective barrier that guards against contaminants and irritants that may delay healing
pH 5.2 to 7.2
Quantity 25 Pk.
Volume (English) 0.112 oz.
Volume (Metric) 3.5 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.