Learn More
Honeywell™ Mounting Brackets for Water-Jel™ Emergency Burn Care Products
Catalog No. 1798571B
Change view
Click to view available options
For Use With (Equipment):
Water-Jel Burn Wrap Canister
Water-Jel Fire Blanket Plus Canister
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
17-985-71B Water-Jel Burn Wrap Canister
17-985-71A Water-Jel Fire Blanket Plus Canister
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17-985-71B Supplier Honeywell™ Supplier No. 049012
Metal mounting brackets for use with Water-Jel™ burn care products.
  • Biodegradable, bacteriostatic and water soluble, Water-Jel immediately cools skin, relieves pain, and protects burn against contamination
  • Ideal first aid treatment for burns, providing emergency care until victim receives medical attention
  • Easy to store, transport, and use—no need for water source or special training
TRUSTED_SUSTAINABILITY
Specifications
Product Type Mounting Brackets
Material Metal
For Use With (Equipment) Water-Jel Burn Wrap Canister
Includes Fasteners