Learn More
Moore Medical Water-Jel™ Triple Antibiotic Ointment
Catalog No. 19898214
Change view
Click to view available options
Volume (Metric):
0.9 g
Catalog No. Volume (Metric)
19-898-214 0.9 g
1 options
This item is not returnable. View return policy
Catalog No. 19-898-214 Supplier Moore Medical Supplier No. 53674
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
View Alternate Product
This item is not returnable. View return policy
Specifications
Product Type Triple Antibiotic Ointment
Active Ingredients Bacitracin Zinc, Neomycin Sulfate, Polymyxin B Sulfate
For Use With (Application) Helps to prevent infection in cuts, scrapes and burns
Quantity 25 Pk.
Volume (English) 0.016 oz.
Volume (Metric) 0.9 g