Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Water, LC-MS Grade, Ricca Chemical
SDP

Catalog No. R91540004C Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
1 L
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R91540004C 4 L
R91540001C 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R91540004C Supplier Ricca Chemical Company Supplier No. R91540004C
Get 2 Extra Waters When You Buy 2
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
IUPAC Name water
SMILES O

Specifications

Absorbance Pass
Color Clear, Colorless
CAS Max % 100
Conductivity 0.056 μS/cm
Percent Purity 100%
Grade LC-MS
Packaging Amber Glass Bottle
Sterility Non-sterile
Quantity 4 L
Chemical Name or Material Water
ShelfLife 18 Months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.