Learn More
Stoelting™ Water Maze
SDP
Catalog No. 10000322 Shop All Stoelting Co Products
Change view
Click to view available options
Animal:
Mouse
Mouse, Rat
Rat
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Animal
10-000-322 Rat
10-000-300 Mouse
10-000-301 Mouse, Rat
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 10-000-322 Supplier Stoelting™ Supplier No. 60235
Add to Cart
Add to Cart
Stoelting™ Water Maze is used to assess spatial and learning memory.
Water Maze tank is filled with water and equipped with a platform which acts as a means of escape for animals. During testing, animals learn the location of the platform using spatial cues.
Specifications
For Use With (Application) Neuroscience
Animal Rat