Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Water, Microbial Cell Culture Grade, Fisher BioReagents

Catalog No. BP28201 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
Packaging:
Nalgene Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BP2820-1 1 L Nalgene Poly Bottle
BP2820-100 100 mL Nalgene Poly Bottle
BP2820-500 500 mL Nalgene Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. BP2820-1
Ultrapure water for preparing microbiological growth media.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.015
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym dihydrogen oxide, dihydrogen monoxide
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name oxidane
SMILES O

Specifications

Color Colorless
Boiling Point 100°C
Conductivity 2μS/cm Max.
Endotoxin 0.025 EU/mL max.
Formula Weight 18.01
Grade Microbial Cell Culture
Packaging Nalgene Poly Bottle
Vapor Pressure 17.5mmHg at 20°C
Filtered Through 0.03 Micron filtered and autoclaved
Density 1.000g/cm³
Quantity 1 L
Physical Form Liquid
Chemical Name or Material Water, 0.03μm filtered and autoclaved
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Statement Low hazard for usual industrial or commercial handling.
Precautionary Statement Emergency Overview
Handle in accordance with good industrial hygiene and safety practice. Rinse with water. Flush eyes with water as a precaution. Move to fresh air. If breathing is difficult, give oxygen. Do not induce vomiting. Obtain medical attention. . . .

NFPA

Health:0
Flammability:0
Instability:0
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.