Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quality Biological Inc Water, Molecular Biology, 4 Liters
SDP

Catalog No. 50146829
Encompass_Preferred

QBI's Molecular Biology Water is prepared by reverse osmosis Passed through fine carbon Deionized through two resin beds Serially filtered through 0.1 µm positively charged membranes

DNase, RNase, and protease tested

Useful in reconstituting nucleic acids and/or proteins, RNA applications (i.e. Preparation of an RNA probe), DNA applications (i.e. Subcloning in pUC plasmids), and protein applications (i.e. Western Blotting). Also, useful as a diluent or solvent.

Storage Conditions: 15 °C to 30 °C
Shipping Condition: ambient temperature
Sterile Filtered & Deionized Tested
DNase, RNase, and Protease Tested
Max Shelf Life: 36 month from date of manufacture

Product and/or packaging contains recycled/renewable content. Product and/or packaging is recyclable or reduces waste. Product and/or packaging is safer to human and environmental health.

Catalog No. 50-146-829 Supplier Quality Biological Inc Supplier No. 351029491
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

QBI's Molecular Biology Water is prepared by reverse osmosis Passed through fine carbon Deionized through two resin beds Serially filtered through 0.1 µm positively charged membranes

DNase, RNase, and protease tested

Useful in reconstituting nucleic acids and/or proteins, RNA applications (i.e. Preparation of an RNA probe), DNA applications (i.e. Subcloning in pUC plasmids), and protein applications (i.e. Western Blotting). Also, useful as a diluent or solvent.

Storage Conditions: 15 °C to 30 °C
Shipping Condition: ambient temperature
Sterile Filtered & Deionized Tested
DNase, RNase, and Protease Tested
Max Shelf Life: 36 month from date of manufacture

Product and/or packaging contains recycled/renewable content. Product and/or packaging is recyclable or reduces waste. Product and/or packaging is safer to human and environmental health.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.