Learn More
Alkali Scientific Water, Molecular Biology Grade,DNase and RNase Free, 10 Liter Cubitainer
SDP
Catalog No. 5000660453
Encompass_Preferred
Water, Molecular Biology Grade, DNase and RNase Free, 10 Liter Cubitainer is a premium water product for molecular biology laboratories, specially purified to remove DNase and RNase to prevent nucleic acid degradation. It is essential for preparing buffers, reagents, and other solutions where contamination with nucleases can lead to inaccurate results. This 10-liter cubitainer is ideal for laboratories that require large quantities of high-purity water. The container is designed to maintain sterility, and the water is suitable for sensitive experiments such as PCR, RNA extractions, and other applications where contaminant-free water is critical.
Catalog No. 50-006-60453 Supplier Alkali Scientific Supplier No. WAT004CP
Water, Molecular Biology Grade, DNase and RNase Free, 10 Liter Cubitainer is a premium water product for molecular biology laboratories, specially purified to remove DNase and RNase to prevent nucleic acid degradation. It is essential for preparing buffers, reagents, and other solutions where contamination with nucleases can lead to inaccurate results. This 10-liter cubitainer is ideal for laboratories that require large quantities of high-purity water. The container is designed to maintain sterility, and the water is suitable for sensitive experiments such as PCR, RNA extractions, and other applications where contaminant-free water is critical.