Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Water Molecular Biology Grade, Lab Chem

Catalog No. LC2672044 Shop All LabChem Products
Change view
Click to view available options
Quantity:
4 x 4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC2672044 4 x 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC2672044 Supplier LabChem, Inc. Supplier No. LC2672044
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

  • Water Molecular Biology Grade

Chemical Identifiers

CAS 7732-18-5
Molecular Weight (g/mol) 18

Specifications

Percent Purity 100%
Grade Molecular Biology
Packaging Plastic Bottle
Quantity 4 x 4 L
Chemical Name or Material Water Molecular Biology Grade
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.