Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Water, Molecular Grade, Vion™
GREENER_CHOICESCI_ED

Catalog No. S01913 Shop All Aldon Products
Change view
Click to view available options
Quantity:
60 mL
125 mL
250 mL
500 mL
1 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S01913 125 mL
S00034 60 mL
S00011 250 mL
S00030 500 mL
S00003 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. S01913 Supplier Aldon Corporation Supplier No. VNWW7003125ML
Add to Cart
Add to Cart

Cell growth and protein expression require the purest water possible to prevent unwanted contaminants. Vion's molecular grade water meets strict specifications to assist in your microbiological applications.

Cell growth and protein expression require the purest water possible to ensure the organism of interest is the only thing growing. Our Molecular Grade Deionized Water is 0.035 micron filtered, UV-sterilized, and tested to ensure maximum suitablility for use in microbiological applications.

Chemical Identifiers

CAS 7732-18-5
Molecular Weight (g/mol) 18.01 g/mol.

Specifications

Percent Purity 100%
Grade Molecular Biology
Packaging PET
Quantity 125 mL
Chemical Name or Material Deionized Water

This substance has not been classified at this time according to the Globally Harmonized System (GHS) of Classification and Labeling of Chemicals.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.