Learn More
Growcells WATER NUCLEASE FREE 10L EA
Catalog No. 501034780
Encompass_Preferred
Molecular Biology Grade/Nuclease Free Water; 18.2 megOhm, 0.1um sterile filtered and autoclaved in polycarbonate bottles.
Catalog No. 50-103-4780 Supplier Growcells Supplier No. NUPW010L
Molecular Biology Grade/Nuclease Free Water; 18.2 megOhm, 0.1um sterile filtered and autoclaved in polycarbonate bottles.