Learn More
Growcells WATER NUCLEASE FREE 125ML PK4
Catalog No. 501034773
Encompass_Preferred
Molecular Biology Grade/Nuclease Free Water; 18.2 megOhm, 0.1um sterile filtered and autoclaved in TriForest polycarbonate bottles.
Catalog No. 50-103-4773 Supplier Growcells Supplier No. NUPW01254
Molecular Biology Grade/Nuclease Free Water; 18.2 megOhm, 0.1um sterile filtered and autoclaved in TriForest polycarbonate bottles.