Learn More
Growcells WATER NUCLEASE FREE 2L EA
Catalog No. 501034778
Encompass_Preferred
Molecular Biology Grade/Nuclease Free Water; 18.2 megOhm, 0.1um sterile filtered and autoclaved in TriForest polycarbonate bottles.
Catalog No. 50-103-4778 Supplier Growcells Supplier No. NUPW2000
Molecular Biology Grade/Nuclease Free Water; 18.2 megOhm, 0.1um sterile filtered and autoclaved in TriForest polycarbonate bottles.