Learn More
McMaster-Carr Water Overflow Buzzer, 96 dB @ 1 ft., includes Hook and Loop Mounting Tape
Catalog No. NC2894965
Encompass
When water contacts the sensor, it triggers a buzzer to alert personnel to a leak or overflow hazard. Frequently used to monitor sinks, pipe lines, tubs, and sump pumps.
Catalog No. NC2894965 Supplier McMaster-Carr Supplier No. 1377T63
When water contacts the sensor, it triggers a buzzer to alert personnel to a leak or overflow hazard. Frequently used to monitor sinks, pipe lines, tubs, and sump pumps.