Learn More
Teknova Water, RNase and DNase Tested, PCR Certified. 100mL, Sterile.
SDP
Catalog No. NC2037137
Encompass
PCR testing is an essential part of quality control for all water products, and the only way to guarantee that a product is safe to use for PCR-based applications. When you choose PCR Certified products, you can be confident that they have undergone a rigorous testing process to ensure that they are free of contamination.
This item is not returnable. View return policy
Catalog No. NC2037137 Supplier Teknova Supplier No. W3335
PCR testing is an essential part of quality control for all water products, and the only way to guarantee that a product is safe to use for PCR-based applications. When you choose PCR Certified products, you can be confident that they have undergone a rigorous testing process to ensure that they are free of contamination.