Learn More
Sigma Aldrich Fine Chemicals Biosciences Water PCR Grade
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 3315959001
Encompass
Water PCR Grade is specially purified double-distilled deionized and autoclaved. The production process ensures that even according to Roche s strict Quality Control procedures the water does not contain detectable amounts of nucleic acid. It is routinely tested in PCR with conventional thermal cyclers and with LightCycler(R) Instruments.
Catalog No. 50-290-0984