Learn More
Grainger Insulated Tools US Water Pump Pliers, 10 in., 1,000 V, Nylon Thermoplastic, 1 ea.
Catalog No. 502779514
Encompass_Preferred
These 1,000 V insulated water pump pliers are designed for live-line working and general maintenance tasks. Manufactured with a durable steel construction and injection moulded with nylon thermoplastic, these groove joint pliers provide reliable performance in demanding environments. The tool is compliant with international safety standards and offers versatile adjustment capabilities for various gripping applications.
  • 6 jaw positions for flexible manual adjustments
  • 1,000 V insulation rating for electrical safety
  • Curved jaw design with a serrated texture for secure gripping
  • Chrome plated finish for corrosion resistance
  • Operational temperature range of -40°C to +70°C
  • Standards compliance: ASTM F1505 and IEC 60900:2012
  • Tether capable design
Catalog No. 50-277-9514 Supplier Grainger Supplier No. 19MU95
These 1,000 V insulated water pump pliers are designed for live-line working and general maintenance tasks. Manufactured with a durable steel construction and injection moulded with nylon thermoplastic, these groove joint pliers provide reliable performance in demanding environments. The tool is compliant with international safety standards and offers versatile adjustment capabilities for various gripping applications.
  • 6 jaw positions for flexible manual adjustments
  • 1,000 V insulation rating for electrical safety
  • Curved jaw design with a serrated texture for secure gripping
  • Chrome plated finish for corrosion resistance
  • Operational temperature range of -40°C to +70°C
  • Standards compliance: ASTM F1505 and IEC 60900:2012
  • Tether capable design