Learn More
Caron Products And Services Pump, Water, 24 VDC, Transfer, Viton O-Rings, 1/4" One Touch Connections
Catalog No. NC2460032
Encompass_Preferred
Pump, Water, 24 VDC, Transfer, Viton O-Rings, 1/4" One Touch Connections
Catalog No. NC2460032 Supplier Caron Products And Services Supplier No. PMP151
Pump, Water, 24 VDC, Transfer, Viton O-Rings, 1/4" One Touch Connections