Learn More
Hach Company Water Purification Cartridge
Supplier:  Hach Company 2967000
Encompass
Water Purification System Adsorber Filtration CartridgeConvenient economical high quality modular water filtration system that operates on city water pressure and can easily be expanded to fit most needs No electricity required Up to 18 Megohm water quality can be achieved with this system Accepts 1/4in tubing Features
Catalog No. 50-002-07010