Learn More
Hach Company Water Purification Univ. Cart.
Supplier:  Hach Company 2967100
Encompass
Water Purification System Universal Filtration CartridgeConvenient economical high quality modular water filtration system that operates on city water pressure and can easily be expanded to fit most needs No electricity required Up to 18 Megohm water quality can be achieved with this system Accepts 1/4in tubing Features
Catalog No. 50-002-06995