Learn More
Yamato Scientific America, Inc. Compact and economical water purification system in 115V with 20L tank that produces Type 1 deionized water and Type 2 distilled water
Catalog No. NC2334073 Shop All Yamato Scientific America Inc. Products
Encompass_Preferred
Yamato’s Auto Still WG205 is an economical multi-purpose benchtop high purity water purification system with 20L storage tank. It is equipped with a pre-treatment cartridge to increase the life span of all consumables. In addition, it displays alerts for consumable replacements and keeps track of replacement history. The WG205 produces ASTM Type 1 deionized water and Type 2 distilled water. *This product requires an ion-exchange cartridge for operation. The item number is NC2135080.
Catalog No. NC2334073 Supplier Yamato Scientific America, Inc. Supplier No. WG205115V
Add to Cart
Add to Cart
Request a Quote
Yamato’s Auto Still WG205 is an economical multi-purpose benchtop high purity water purification system with 20L storage tank. It is equipped with a pre-treatment cartridge to increase the life span of all consumables. In addition, it displays alerts for consumable replacements and keeps track of replacement history. The WG205 produces ASTM Type 1 deionized water and Type 2 distilled water. *This product requires an ion-exchange cartridge for operation. The item number is NC2135080.