Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LaMotte™ Water Quality Educator and Monitoring Outfit
SDPSCI_ED

Catalog No. S66223 Shop All LaMotte Company Products
Change view
Click to view available options
Type:
Reagent Refill
Water Quality Educator and Monitoring Outfit
Includes:
Set of refill reagents as originally supplied in the complete kit
pH, nitrate-nitrogen, phosphate, dissolved oxygen, alkalinity, total, turbidity, temperature test factor, The Monitor's Handbook, 100 tests for pH, unlimited temperature tests, aquatic macro invertebrate bioassessment
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Includes
S66223 Water Quality Educator and Monitoring Outfit pH, nitrate-nitrogen, phosphate, dissolved oxygen, alkalinity, total, turbidity, temperature test factor, The Monitor's Handbook, 100 tests for pH, unlimited temperature tests, aquatic macro invertebrate bioassessment
S66224 Reagent Refill Set of refill reagents as originally supplied in the complete kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S66223 Supplier LaMotte™ Supplier No. 587001
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

  • The Monitor's Handbook (71 pages)
  • 100 tests for pH
  • 50 tests for the following five factors: nitrate-nitrogen, phosphate, dissolved oxygen, total alkalinity, turbidity, unlimited temperature tests, aquatic macroinvertebrate bioassessment
  • Lesson plans for grades 5 to 8 and 9 to 12

Ideal for long-term water-quality monitoring.

  • Tests for seven basic water-quality test factors
  • Prepares students for field tests or classroom work
  • Set up a water-quality monitoring program, procure measurements, and interpret results
  • Soft-sided tote made of durable fabric, solid rubber bottom ready for field work; handy side pockets hold thermometer, pens, writing tablets and other tools

Test Factor / Range / Individual Kit Code

  • pH, pH 3.0-10.5 (100 tests), 5858-01
  • Nitrate-Nitrogen, 0-15 ppm (50 tests), 3354-01
  • Phosphate, 0-2.0 ppm (50 tests), 3121-02
  • Dissolved Oxygen, 0-10.0 ppm (50 tests), 5860-01
  • Alkalinity (tot.), 0-200 ppm (50 tests), 4491-DR-01
  • Turbidity, 0-200 JTU (50 tests), 7519-01
  • Temperature, -5° to 45°C, 1066

Specifications

For Use With (Application) Long term water quality monitoring
Grade 5+
Includes pH, nitrate-nitrogen, phosphate, dissolved oxygen, alkalinity, total, turbidity, temperature test factor, The Monitor's Handbook, 100 tests for pH, unlimited temperature tests, aquatic macro invertebrate bioassessment
No. of Tests 50
Type Water Quality Educator and Monitoring Outfit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.