Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Water, Reagent Grade; OmniSolv™, MilliporeSigma™

Catalog No. MWX0004NP20 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
20 L
Packaging:
Plastic Barrel
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MWX0004NP20 20 L Plastic Barrel
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MWX0004NP20 Supplier MilliporeSigma Supplier No. US1WX0004/NP20
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

  • Used for testing USP/NF monographs
  • For HPLC Gradient Analysis, Spectrophotometry, Gas Chromatography
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.015
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name oxidane
SMILES O

Specifications

pH Neutral
Melting Point 0°C
Boiling Point 100°C (1013 hPa)
Formula Weight 18.02g/mol
Packaging Plastic Barrel
Quantity 20 L
Physical Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.