Learn More
Labtech Inc Recirculating Water Chiller 3000w
Catalog No. 501959347
Encompass_Preferred
Recirculating Water Chiller by LabTech, cooling capacity of 3000 Watt at 25˚C, 230V, 8C ~ 35˚C, model H150-3000
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Catalog No. 50-195-9347 Supplier Labtech Inc Supplier No. LW3000
Add to Cart
Add to Cart
Request a Quote
Recirculating Water Chiller by LabTech, cooling capacity of 3000 Watt at 25˚C, 230V, 8C ~ 35˚C, model H150-3000
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.