Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mopec Water Recovery Body Bag
SDP

Catalog No. 22444357 Shop All Mopec Products
Change view
Click to view available options
Dimensions (L x W):
96 x 36in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
22-444-357 96 x 36in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-444-357 Supplier Mopec Supplier No. BE085
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mopec Water Recovery Body Bag with envelope zipper is constructed to meet U.S. military, coast guard, fire department and police dive team standards for use during water recovery of human remains

  • Yellow color allows for high visibility
  • Poly mesh material allows for fast drainage
  • Ten (10) strong, wide web handles
  • Meets U.S. standards for water recovery of human remains
  • Impervious to blood, body fats and other normal body fluids

Specifications

Description 76in. Girth
Closure Type Zip Closure
Color Yellow
For Use With (Application) Water recovery
Material Poly Mesh
Quantity 1
Dimensions (L x W) 96 x 36in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.