Learn More
Thermco Products Inc WATER RESIST POCKET 4 1/2IN
Catalog No. NC3445995
Encompass_Preferred
WATER RESISTANT POCKET 4 1/2″ Stem Digital ThermometersRange: -50°/150°C (-58°/302°F)4 1/2” Stainless Steel Stem w/Probe CoverMin – Max & Hold FunctionWater ResistantNIST Traceable w/CertificateC/F SwitchableResolution: 0.1ºAccuracy: ± 1.0ºC between -20º to 120ºC
Catalog No. NC3445995 Supplier Thermco Products Inc Supplier No. ACC600DIG
WATER RESISTANT POCKET 4 1/2″ Stem Digital ThermometersRange: -50°/150°C (-58°/302°F)4 1/2” Stainless Steel Stem w/Probe CoverMin – Max & Hold FunctionWater ResistantNIST Traceable w/CertificateC/F SwitchableResolution: 0.1ºAccuracy: ± 1.0ºC between -20º to 120ºC