Learn More
Cole-Parmer Tempshield WRL Water-resistant gloves, 12"L, large, 1 pair
Catalog No. NC9942951 Shop All Cole-Parmer Products
Encompass_Preferred
These water-resistant glovesprovide protection in closed cryogenic systems with limited risk of exposure to cryogenic fluids. A stay-dry lining keeps you warm and comfortable. The gloves can not be immersed in liquid nitrogen.
This item is not returnable. View return policy
Catalog No. NC9942951 Supplier Cole-Parmer Supplier No. 0911304
These water-resistant glovesprovide protection in closed cryogenic systems with limited risk of exposure to cryogenic fluids. A stay-dry lining keeps you warm and comfortable. The gloves can not be immersed in liquid nitrogen.