Learn More
Bioworld WATER RNA/DNA PURIFICATION
Supplier:  Bioworld 107601321
Encompass_Preferred
WATER RNA/DNA PURIFICATION KIT- 0.22M: For convenient purification of RNA and DNA from microorganisms in water sample Isolate total DNA and RNA from all microorganisms found in water, including bacteria, fungi and algae; RNA and DNA are ...
Catalog No. 50-196-989