Learn More
Water Rocket and Class Packs
SCI_ED
Catalog No. S06877
Change view
Click to view available options
Product Type:
Water Rocket Class Pack
Water Rocket Single Kit
Includes:
1 Molded nozzle, 1 die cut fin set, 1 soft Foam bumper, assembly instructions
25 Precut rigid Foam fin set, 25 translucent molded nozzles, 25 Foam recovery bumpers, color assembly and use instructions
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Includes
S06877 Water Rocket Single Kit 1 Molded nozzle, 1 die cut fin set, 1 soft Foam bumper, assembly instructions
S06873 Water Rocket Class Pack 25 Precut rigid Foam fin set, 25 translucent molded nozzles, 25 Foam recovery bumpers, color assembly and use instructions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S06877 Supplier Quest Aerospace Inc Supplier No. 7332
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
Requires
Standard bicycle pump with a pressure gauge; assembly of foam fins requires low temperature hot glue gun
Individual components to build a complete water rocket.
Packs consist of:
  • Pre-cut rigid foam fin sets
  • Translucent molded nozzles
  • Foam recovery bumpers
  • Color assembly and use instructions
Specifications
Product Type Water Rocket Single Kit
Includes 1 Molded nozzle, 1 die cut fin set, 1 soft Foam bumper, assembly instructions
For Use With (Application) Perfect low cost flying rocket without the hassles of traditional solid propellant rockets! Theres no fire danger (the rocket fuel is water) and no HazMat fees to ship them!
Subject Physics