Learn More
MilliporeSigma™ Water Sensor
Catalog No. TANKLK001 Shop All MilliporeSigma Products
Change view
Click to view available options
Type:
Water Sensor
Catalog No. Type
TANKLK001 Water Sensor
1 options
This item is not returnable. View return policy
Catalog No. TANKLK001 Supplier MilliporeSigma™ Supplier No. TANKLK001
Add to Cart
Add to Cart
This item is not returnable. View return policy
MilliporeSigma Water Sensor designed to reduce the risk of flooding.
  • Reduces the risk of flooding: no damage to the lab or other instruments.
  • Cable length: 4 meters.
TRUSTED_SUSTAINABILITY
Specifications
Type Water Sensor
For Use With (Equipment) RiOs™ and Elix™ Water Purification Systems