Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Water Soluble Paper Labels with Ribbon

Catalog No. 19000760 Shop All Brady Corporation Products
Change view
Click to view available options
Color:
Black on White
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-000-760 Black on White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-000-760 Supplier Brady™ Supplier No. M21500403
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For the ultimate solution in label and adhesive removal, choose Brady’s B-403 Water Dissolvable Label. Ideal for laboratories, temporary identification needs, work-in-process product flows, and other general identification applications.

  • For the ultimate solution in label and adhesive removal, choose Brady's B-403 Water Dissolvable Label.
  • Ideal for laboratories, temporary identification needs, work-in-process product flows, and other general identification applications.
  • Work-In-Process: warm water and 30 seconds is all it takes to completely remove the label and adhesive.
  • Application Versatility: From laboratories to restaurants, this dissolvable paper material is an excellent solution for temporary identification of any kind.
  • Black print on white paper with easy to apply pressure-sensitive adhesive.
  • Certifications: RoHS compliant to 2011/65/EU (Pass 2015/863).
  • For material specific performance and properties, please refer to the Technical Data Sheet (TDS).

Specifications

Adhesive Type Acrylic
Color Black on White
Finish Matte
Material Paper
Print Technology Thermal Transfer
Product Type Label
Sterility Non-sterile
For Use With (Equipment) Brady Label Printers, Brady M210-LAB Label Printer, Brady M211 Label Printer, BMP21-PLUS Label Printer, BMP21 LAB Label Printer
Length (English) 25 ft.
Width (English) 2.69 in.
Alternate Part Number M21-500-403
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.